Learn More
Eppendorf™ Rotor for Centrifuge 5427 R
Eppendorf Rotor for Centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
Brand: Eppendorf™ 5409704001
886.33 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
- Aerosol-tight options available.
Specifications
Aerosol-tight Rotor | |
19,090 x g | |
Centrifuge 5427 R | |
24 |
24 Spin Columns | |
13,200 rpm | |
Aerosol-tight with Rotor Lid | |
FA-45-24-11-Kit |