missing translation for 'onlineSavingsMsg'
Learn More

BNIP3 Mouse anti-Human, Clone: 2H7D8, Proteintech

Mouse Monoclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen BNIP3
Clone 2H7D8
Concentration 1000 μg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17679776
View Documents
Proteintech
68091-1-IG-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17669776
View Documents
Proteintech
68091-1-IG-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BNIP3 Monoclonal antibody apecifically detetects BNIP3 antigen in Human Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

BNIP3
1000 μg/mL
Monoclonal
Liquid
Human
BC009342
BNIP3
IgG2b
Protein A purification
2H7D8
Western Blot, ELISA
Unconjugated
Mouse
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
664
BNIP3 fusion protein Ag27935
Primary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.