missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIP3 Mouse anti-Human, Clone: 2H7D8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68091-1-IG-20UL
This item is not returnable.
View return policy
Description
BNIP3 Monoclonal antibody apecifically detetects BNIP3 antigen in Human Samples. It is validated for Western Blot, ELISA.Specifications
| BNIP3 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| BNIP3 | |
| BNIP3 fusion protein Ag27935 | |
| 20 μL | |
| 664 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG2b |
| Western Blot, ELISA | |
| 2H7D8 | |
| Unconjugated | |
| BC009342 | |
| Mouse | |
| Protein A purification | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction