Learn More
B Braun™ Biemer Vessel Clips
For temporary occlusion in surgical applications.
Brand:  B Braun™ FD554R
 View more versions of this product
Product Code. 10147954
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Blade gripping surface with oblique serrations for safe but careful gripping of vessels
  • No risk due to rigidly fixed pressure measure which guarantees application without any risk
  • Closing force is measured at 1/3 of the blades at 1 mm opening
Specifications
Specifications
Biemer Forceps
Stainless Steel
4 mm opening
1.4 mm (Jaw)
15 to 20 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers