Learn More
Axygen™ Maximum Recovery™ Polypropylene Pipette Tip
For ultra-low retention and high yield
Brand:  Axygen™ T-200-Y-R
 View more versions of this product
Product Code. 12773109
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Beveled
  • Maxymum Recovery technology for ultra-low retention and high yield
  • Sterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 μL
Yellow
Used with single and multichannel pipettes
Universal Fit
Polypropylene
Sterile
DNase, RNase, and Pyrogen Free
Rack
Maxymum Recovery
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ Maximum Recovery™ Polypropylene Pipette Tip > Racked; Standard; 1-200 μL