Learn More
Abeba™ Arrow 38202 Shoes
Antistatic with heel patch microfiber. Sanitized™ treated.
Brand: Abeba™ 38202/50
This item is not returnable.
View return policy
Description
- CE EN ISO 20347:2012 OB A, FO SRA
- Smooth leather white
- Slip resistance PU/PU outsole sole
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
- Microfiber heel patch
- Lining microfiber, respiration absorbing
Specifications
CE EN ISO 20347:2012 OB A, FO SRA | |
Microfiber | |
50 | |
Polyurethane |
White | |
Leather | |
Slip-resistant | |
38202 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Abeba™ Arrow 38202 Shoes > Size: 50