Learn More
ARN2966, MedChemExpress
MedChemExpress ARN2966 is a potent post-transcriptional modulator of APP expression; reduces expression of APP with resultant lower production of A(beta).
Brand:  Medchem Express HY-18292
Encompass
 View more versions of this product
Product Code. 16439977
  • € 151.50 / 5mg
Explore more special offers
Description
Description
ARN2966 is a potent post-transcriptional modulator of APP expression
Specifications
Specifications
ARN2966
Yellow
C12H12N2O
DMSO : 110 mg/mL (549.34 mM; Need ultrasonic)
200.24
98.0%
Solid
102212-26-0
Research
5 mg
OC1=CC=CC=C1NCC2=NC=CC=C2
200.24
Research
Neuroscience-Neurodegeneration
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only