Learn More
ARN2966, MedChemExpress
MedChemExpress ARN2966 is a potent post-transcriptional modulator of APP expression; reduces expression of APP with resultant lower production of A(beta).
Brand: Medchem Express HY-18292
Description
ARN2966 is a potent post-transcriptional modulator of APP expressionSpecifications
ARN2966 | |
Yellow | |
C12H12N2O | |
DMSO : 110 mg/mL (549.34 mM; Need ultrasonic) | |
200.24 | |
98.0% | |
Solid |
102212-26-0 | |
Research | |
5 mg | |
OC1=CC=CC=C1NCC2=NC=CC=C2 | |
200.24 | |
Research | |
Neuroscience-Neurodegeneration |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only