Learn More
Anethole, MedChemExpress
MedChemExpress Anethole is a type of aromatic compound that occurs widely in nature, widely used as a flavoring substance.
Brand:  Medchem Express HY-B0900
Encompass
 View more versions of this product
Product Code. 16432226
  • € 111.00 / 500mg
Explore more special offers
Description
Description
Anethole is a type of aromatic compound that occurs widely in nature, widely used as a flavoring substance.
Specifications
Specifications
Anethole
White
C10H12O
Anise camphor p-Propenylanisole Isoestragole
C/C=C/C1=CC=C(OC)C=C1
148.2
Research
Cancer-programmed cell death
104-46-1
Research
500 mg
DMSO : ≥ 100 mg/mL (674.76 mM)
148.2
98.46%
Solid-Liquid Mixture
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Anethole, MedChemExpress
Research purposes only