Learn More
Fisherbrand™ Aluminum Gates Gel Casting Tray
Ideal for standard preparatory, analytical electrophoresis techniques
€ 103.00 - € 180.00
Product Code | Brand | Length (Metric) Plate | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) Plate | Width (Metric) | Price | Quantity & Availability | ||||
11716165
|
Fisherbrand™
SVGGT7 |
70 mm | 70 mm | N/A | |||||
11726155
|
Fisherbrand™
VGGT10 |
100 mm | 150 mm | N/A | |||||
11746155
|
Fisherbrand™
VGGT20 |
200 mm | 150 mm | N/A | |||||
11716155
|
Fisherbrand™
VGGT7 |
70 mm | 150 mm | N/A | |||||
Description
- Three different Vari-gel tanks, 150mm, 100mm and 70mm are available accommodating up to four different gel tray sizes
- UV transparent gel trays allow for placement directly onto a UV transilluminator for gel viewing once stained
- Asymmetric electrode offset - prevents accidental polarity reversal ensuring no possibility of sample loss into the buffer
- aluminum casting gates - ensure easy casting of the gel inside or outside of the tank. They slide into pre-formed grooves at the end of the gel tray providing a secure seal against gel leakage
- Different gel sizes can be interchanged in one tank, combining superb resolution with ease of use.
Specifications
Gel Casting Tray | |
Aluminum |