Learn More
Fisherbrand™ Aluminum Gates Gel Casting Tray
Ideal for standard preparatory, analytical electrophoresis techniques
Brand: Fisherbrand™ VGGT7
This item is not returnable.
View return policy
Description
- UV transparent
- For visualisation with fluorescent numbering for SNP recognition, traceability
- Accessory for Vari-gel horizontal gel electrophoresis systems
Specifications
Gel Casting Tray | |
Aluminum | |
70 mm | |
150 mm |