Learn More
Fisherbrand™ Aluminum Gates Gel Casting Tray
Ideal for standard preparatory, analytical electrophoresis techniques
Brand:  Fisherbrand™ VGGT7
 View more versions of this product
Product Code. 11716155
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • UV transparent
  • For visualisation with fluorescent numbering for SNP recognition, traceability
  • Accessory for Vari-gel horizontal gel electrophoresis systems
Specifications
Specifications
Gel Casting Tray
Aluminum
70 mm
150 mm