Learn More
A939572, MedChemExpress
MedChemExpress A939572 is a potent, and orally bioavailable stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50 values of mSCD1 and hSCD1, respectively.
Brand: Medchem Express HY-50709

Description
A939572 is a potent, and orally bioavailable stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50 values of mSCD1 and hSCD1, respectively.Specifications
A939572 | |
White | |
C20H22ClN3O3 | |
DMSO : 100 mg/mL (257.82 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) | |
387.86 | |
99.54% | |
Solid |
1032229-33-6 | |
Research | |
5 mg | |
O=C(N1CCC(OC2=C(Cl)C=CC=C2)CC1)NC3=CC=CC(C(NC)=O)=C3 | |
387.86 | |
Research | |
Cancer-programmed cell death |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only