missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A01

High affinity Smurf1 inhibitor; enhances BMP signaling

€ 228.00 - € 872.00

Specifications

Purity >98%
Product Type A01
Molecular Weight (g/mol) 512.93
Inhibitors A01
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18732459
View Documents
Tocris Bioscience™
5397/10
10 mg
€ 228.00
10mg
Please to purchase this item. Need a web account? Register with us today!
18767078
View Documents
Tocris Bioscience™
5397/50
50 mg
€ 872.00
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

High affinity Smurf1 E3 ubiquitin ligase inhibitor (Kd = 3.7 nM). Attenuates Smurf1-mediated Smad 1/5 degradation and enhances BMP signaling. Potentiates BMP-2-induced osteoblastic activity in C2C12 myoblasts and MC3T3-E1 osteoblast precursor cells.
Specifications

Specifications

>98%
A01
512.93
A01
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only