missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Solid Bottomed Microplates
For high-throughput screening and assay
€ 132.00
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 8 x 12 |
Color | Clear |
Material | Polystyrene |
Product Code | Brand | Well Shape | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Well Shape | Price | Quantity & Availability | |||||
10625821
|
Greiner Bio-One
650161 |
Round Bottom (U-well) |
€ 132.00
Pack of 100 |
N/A | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Ideally suited for agglutination tests
- No sharp corners, which means that they can be pipetted easily and cleanly
- Suitable for +/- analyses
- Solid bot
Specifications
Standard | |
8 x 12 | |
Polystyrene | |
Sterile | |
4800 x g (swinging bucket rotor) | |
100/Cs. |
Without Lid | |
Clear | |
96 | |
Non-treated | |
100 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title