missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Solid Bottomed Microplates
For high-throughput screening and assay
Brand: Greiner Bio-One 650161
This item is not returnable.
View return policy
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Ideally suited for agglutination tests
- No sharp corners, which means that they can be pipetted easily and cleanly
- Suitable for +/- analyses
- Solid bot
Specifications
No | |
Without Lid | |
Non Binding | |
Polystyrene | |
40 to 280 μL | |
Non-treated | |
Round Bottom (U-well) | |
100 | |
Clear |
Standard | |
8 x 12 | |
Clear | |
96 | |
Sterile | |
323 μL | |
4800 x g (swinging bucket rotor) | |
100/Cs. | |
Chimney |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction