Learn More
5-(Trifluoromethoxy)salicylaldehyde, 98+%, Thermo Scientific Chemicals
Product Code. 11373966
Click to view available options
Quantity:
250 mg
1 g
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 93249-62-8
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD00075249
InChI Key WQUZBERVMUEJTD-UHFFFAOYSA-N
Synonym 2-hydroxy-5-trifluoromethoxy benzaldehyde, 5-trifluoromethoxy salicylaldehyde, benzaldehyde, 2-hydroxy-5-trifluoromethoxy, acmc-20antc, 5-trifluoromethoxysalicylaldehyde, 5-trifluoromethoxy-salicylaldehyde, 2-hydroxy-5-trifluoromethoxy-benzaldehyde, 20hydroxy-5-trifluoromethoxy benzaldehyde, 2-oxidanyl-5-trifluoromethyloxy benzaldehyde, benzaldehyde,2-hydroxy-5-trifluoromethoxy
PubChem CID 1268058
IUPAC Name 2-hydroxy-5-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=C(C=C1OC(F)(F)F)C=O)O
Specifications
Melting Point 31°C to 33°C
Boiling Point 82°C (60 mmHg)
Flash Point >110°C (230°F)
Quantity 250 mg
Beilstein 7023825
Sensitivity Air Sensitive
Formula Weight 206.12
Percent Purity ≥98%
Chemical Name or Material 5-(Trifluoromethoxy)salicylaldehyde