Learn More
4-Methoxyphenylboronic acid, 97%, Thermo Scientific Chemicals
€ 44.82 - € 328.64
Chemical Identifiers
CAS | 5720-07-0 |
---|---|
Molecular Formula | C7H9BO3 |
Molecular Weight (g/mol) | 151.96 |
MDL Number | MFCD00039139 |
InChI Key | VOAAEKKFGLPLLU-UHFFFAOYSA-N |
Synonym | 4-methoxyphenyl boronic acid, 4-methoxybenzeneboronic acid, p-anisylboronic acid, p-methoxyphenylboronic acid, p-methoxybenzeneboronic acid, 4-methoxyphenyl boranediol, benzeneboronic acid, p-methoxy, 4-boronoanisole, 4-methoxyphenylboronicacidShow More |
PubChem CID | 201262 |
IUPAC Name | (4-methoxyphenyl)boronic acid |
SMILES | COC1=CC=C(C=C1)B(O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10328790
|
Thermo Scientific Acros
309480010 |
1 g | Glass bottle |
€ 44.82
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10415365
|
Thermo Scientific Acros
309480050 |
5 g | Glass bottle |
€ 165.83
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10297343
|
Thermo Scientific Acros
309480250 |
25 g | Glass bottle |
€ 328.64
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5720-07-0 | |
151.96 | |
VOAAEKKFGLPLLU-UHFFFAOYSA-N | |
201262 | |
COC1=CC=C(C=C1)B(O)O |
C7H9BO3 | |
MFCD00039139 | |
4-methoxyphenyl boronic acid, 4-methoxybenzeneboronic acid, p-anisylboronic acid, p-methoxyphenylboronic acid, p-methoxybenzeneboronic acid, 4-methoxyphenyl boranediol, benzeneboronic acid, p-methoxy, 4-boronoanisole, 4-methoxyphenylboronicacidShow More | |
(4-methoxyphenyl)boronic acid |
Specifications
5720-07-0 | |
100.0 | |
White to Beige | |
97% | |
CH3OC6H4B(OH)2 | |
4-methoxyphenyl boronic acid, 4-methoxybenzeneboronic acid, p-anisylboronic acid, p-methoxyphenylboronic acid, p-methoxybenzeneboronic acid, 4-methoxyphenyl boranediol, benzeneboronic acid, p-methoxy, 4-boronoanisole, 4-methoxyphenylboronicacid | |
COC1=CC=C(C=C1)B(O)O | |
151.96 | |
151.96 | |
Crystalline Powder |
96.0 | |
208.0°C to 210.0°C | |
Authentic | |
C7H9BO3 | |
MFCD00039139 | |
VOAAEKKFGLPLLU-UHFFFAOYSA-N | |
(4-methoxyphenyl)boronic acid | |
201262 | |
97% | |
4-Methoxyphenylboronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
GHS Signal Word: Warning
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.