missing translation for 'onlineSavingsMsg'
Learn More
4-Methoxyphenylboronic acid, 97%, Thermo Scientific Chemicals Product Code.: 10297343

4-Methoxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 10297343
25 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10297343

Brand: Thermo Scientific Acros 309480250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5720-07-0
Molecular Formula C7H9BO3
Molecular Weight (g/mol) 151.96
MDL Number MFCD00039139
InChI Key VOAAEKKFGLPLLU-UHFFFAOYSA-N
Synonym 4-methoxyphenyl boronic acid, 4-methoxybenzeneboronic acid, p-anisylboronic acid, p-methoxyphenylboronic acid, p-methoxybenzeneboronic acid, 4-methoxyphenyl boranediol, benzeneboronic acid, p-methoxy, 4-boronoanisole, 4-methoxyphenylboronicacidShow MoreShow Less
PubChem CID 201262
IUPAC Name (4-methoxyphenyl)boronic acid
SMILES COC1=CC=C(C=C1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 208.0°C to 210.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H4B(OH)2
Quantity 25 g
Formula Weight 151.96
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenylboronic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0432166 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog Number309480250,309480010,309480050
Lot NumberA0402909 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number309480250,309480010,309480050
Lot NumberA0399954 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number309480250,309480010,309480050
Lot NumberA0414842 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number309480250,309480010,309480050
compliance-icons
Product Identifier
  • 4-Methoxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 4-Methoxyphenylboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methoxyphenylboronic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.