missing translation for 'onlineSavingsMsg'
Learn More
4-Dimethylaminobenzaldehyde, 98%, Thermo Scientific Chemicals Product Code.: 10306650

4-Dimethylaminobenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 10306650
500 g
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy

Product Code. 10306650

Brand: Thermo Scientific Acros 149285000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70°C to 75°C
Color White to Off-white
Density 5.15
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula (CH3)2NC6H4CHO
Quantity 500 g
Beilstein 14, 31
Fieser 01,273; 02,146
Merck Index 15, 3252
Formula Weight 149.19
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminobenzaldehyde, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Dimethylaminobenzaldehyde, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.