Learn More
4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals Product Code.: 11424174
4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals
Product Code. 11424174
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
Product Code. 11424174
Brand: Thermo Scientific Alfa Aesar A12518.22
Request Bulk or Custom Format
4-Bromobenzoic acid was used to study the metabolic fate of 2-,3-and 4-bromo benzoic acids in rat hepatocytes incubation using high temperature liquid chromatography. It was used in bromine-specific detection of the metabolites of 2-,3-and 4-bromobenzoic acid in the urine and bile of rats by inductively coupled plasma mass spectrometry. It is also used as an intermediate for the synthesis of agrochemicals, pharmaceutical, chemical intermediates and liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromobenzoic acid was used to study the metabolic fate of 2-,3-and 4-bromo benzoic acids in rat hepatocytes incubation using high temperature liquid chromatography. It was used in bromine-specific detection of the metabolites of 2-,3-and 4-bromobenzoic acid in the urine and bile of rats by inductively coupled plasma mass spectrometry. It is also used as an intermediate for the synthesis of agrochemicals, pharmaceutical, chemical intermediates and liquid crystals.
Solubility
Slightly soluble in hot water. Soluble in alcohol and ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 586-76-5
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD00002529
InChI Key TUXYZHVUPGXXQG-UHFFFAOYSA-N
Synonym p-bromobenzoic acid, benzoic acid, 4-bromo, p-carboxybromobenzene, benzoic acid, p-bromo, p-bromobenzenecarboxylic acid, 4-bromo-benzoic acid, 4-brom-benzoesaeure, 4-carboxybromobenzene, para bromo benzoic acid, 4bromo-benzoic acidShow MoreShow Less
PubChem CID 11464
ChEBI CHEBI:60698
IUPAC Name 4-bromobenzoic acid
SMILES OC(=O)C1=CC=C(Br)C=C1
Specifications
Melting Point 253°C to 258°C
Density 1.894
Odor Odorless
Quantity 100 g
Beilstein 1906923
Merck Index 14,1408
Solubility Information Slightly soluble in hot water. Soluble in alcohol and ether.
Formula Weight 201.02
Percent Purity ≥98%
Chemical Name or Material 4-Bromobenzoic acid
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252255 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog NumberA12518.14,A12518.36,A12518.22
Lot Number10249589 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberA12518.14,A12518.36,A12518.22
Lot Number10248925 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog NumberA12518.14,A12518.36,A12518.22
Lot Number10244863 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA12518.14,A12518.36,A12518.22
Lot Number10193296 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog NumberA12518.14,A12518.36,A12518.22
compliance-icons
Product Identifier
  • 4-Bromobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contains : 4-Bromobenzoic acid
RUO – Research Use Only
Product Content Correction
Product Title
4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals >