Learn More
3-(Phenoxymethyl)benzoic acid, 97%, Thermo Scientific™
Product Code. p-4391444
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
Product Code. 10316474
Brand: Thermo Scientific Maybridge CC63601CB
Request Bulk or Custom Format
Chemical Identifiers
CAS 31719-75-2
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
InChI Key URLYREZIPSJJQU-UHFFFAOYSA-N
Synonym 3-phenoxymethyl benzoic acid, benzoic acid,3-phenoxymethyl, 4rg
PubChem CID 3729884
IUPAC Name 3-(phenoxymethyl)benzoic acid
SMILES C1=CC=C(C=C1)OCC2=CC=CC(=C2)C(=O)O
Specifications
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 228.25
Percent Purity 97%
Chemical Name or Material 3-(phenoxymethyl)benzoic acid