missing translation for 'onlineSavingsMsg'
Learn More
3-Hydroxybenzoic acid, 99%, Thermo Scientific Chemicals Product Code.: 11400204

3-Hydroxybenzoic acid, 99%, Thermo Scientific Chemicals

Product Code. 11400204
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11400204

Brand: Thermo Scientific Alfa Aesar A13628.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Hydroxybenzoic acid is used in perfumery. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybenzoic acid is used in perfumery. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-06-9
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002506
InChI Key IJFXRHURBJZNAO-UHFFFAOYSA-N
Synonym m-hydroxybenzoic acid, m-salicylic acid, 3-carboxyphenol, benzoic acid, 3-hydroxy, m-hba, 3-hydroxybenzoate, benzoic acid, m-hydroxy, acido m-idrossibenzoico, meta-hydroxybenzoic acid, kyselina 3-hydroxybenzoovaShow MoreShow Less
PubChem CID 7420
ChEBI CHEBI:30764
IUPAC Name 3-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC(O)=C1

Specifications

Melting Point 201°C to 203°C
Flash Point 220°C (428°F)
Odor Odorless
Quantity 500 g
Beilstein 508160
Solubility Information Slightly soluble in water.
Formula Weight 138.12
Percent Purity 99%
Chemical Name or Material 3-Hydroxybenzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0447141 Certificate TypeCertificate of Analysis Date03/01/2024 Catalog NumberA13628.36,A13628.22
Lot Number10229965 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13628.36,A13628.22
Lot Number10229966 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13628.36,A13628.22
Lot Number10206594 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13628.36,A13628.22
Lot Number10174131 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13628.36,A13628.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Hydroxybenzoic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.