missing translation for 'onlineSavingsMsg'
Learn More

3-Aminomethyl-benzoic acid methyl ester hydrochloride, 95%, Thermo Scientific™

Product Code. p-4510418
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 10300593

Brand: Thermo Scientific Maybridge MO07352CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17841-68-8
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
InChI Key UOWRPTFJISFGPI-UHFFFAOYSA-N
Synonym methyl 3-aminomethyl benzoate hydrochloride, 3-aminomethylbenzoic acid methyl ester hydrochloride, 3-aminomethyl-benzoic acid methyl ester hydrochloride, methyl 3-aminomethylbenzoate hcl, methyl 3-amino methyl benzoate hydrochloride, methyl 3-aminomethylbenzoate hydrochloride, methyl 3-aminomethyl benzoate hcl, benzoic acid, 3-aminomethyl-, methyl ester, hydrochloride, methyl 3-aminomethyl benzoate, chloride, pubchem11178
PubChem CID 2794825
IUPAC Name methyl 3-(aminomethyl)benzoate;hydrochloride
SMILES COC(=O)C1=CC(=CC=C1)CN.Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 201.65
Percent Purity 95%
Chemical Name or Material 3-Aminomethylbenzoic acid methyl ester hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.