Learn More
3,5-Difluoro-2-methylbenzaldehyde, 97%, Thermo Scientific™
Product Code. 15494497
Click to view available options
Quantity:
250 mg
1 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1017778-68-5
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.132
MDL Number MFCD09832335
InChI Key MTPAERCJUJOIAH-UHFFFAOYSA-N
Synonym 3,5-difluoro-2-methylbenzaldehyde, jrd
PubChem CID 20112146
IUPAC Name 3,5-difluoro-2-methylbenzaldehyde
SMILES CC1=C(C=C(C=C1C=O)F)F
Specifications
Melting Point 32°C to 35°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 156.13
Percent Purity 97%
Chemical Name or Material 3,5-Difluoro-2-methylbenzaldehyde