missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromo-o-phenylenediamine monohydrochloride, 99%, Thermo Scientific™
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Specifications
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 238°C |
Color | Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Linear Formula | Br2C6H2(NH2)2·HCl |
Quantity | 100 mg |
Beilstein | 13, 29 |
Show More |
Safety and Handling
Product Identifier
- 3,5-Dibromo-o-phenylenediamine monohydrochloride
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'