missing translation for 'onlineSavingsMsg'
Learn More

(2-Methoxy-5-nitrophenyl)acetic acid, 97%, Thermo Scientific Chemicals

Product Code. 15487317
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51073-04-2
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.17
MDL Number MFCD02664832
InChI Key TZQZNDOEOCOJFS-UHFFFAOYSA-N
Synonym 2-2-methoxy-5-nitrophenyl acetic acid, 2-methoxy-5-nitrophenyl acetic acid, 2-methoxy-5-nitrophenylacetic acid, benzeneacetic acid, 2-methoxy-5-nitro, 2-methoxy-5-nitro-phenyl-acetic acid
PubChem CID 3519298
IUPAC Name 2-(2-methoxy-5-nitrophenyl)acetic acid
SMILES COC1=C(CC(O)=O)C=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 153°C to 155°C
Quantity 5 g
Formula Weight 211.17
Percent Purity 97%
Chemical Name or Material (2-Methoxy-5-nitrophenyl)acetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'