Learn More
Hamilton™ 2 μL 7000 Series Microliter Syringe
The 7000 series syringes are modified Microliter syringes with a plunger-in-needle design.
Brand:  Hamilton™ CAL88511
 View more versions of this product
Product Code. 17864222
  • € 350.00 / Each
Explore more special offers
Description
Description
  • The only zero-dead volume syringe available.
  • The fine gauge tungsten plunger wire extends to the tip of the needle, ensuring that all solvent is expelled, dispensing volumes up to 2 μL.
Specifications
Specifications
Autoclavable
23 G
0.0081 in.
0.020 in.
0.206 mm
0.515 mm
Microliter Syringe
115°C
7002 KH
1
69.9 mm
Knurled Hub
Borosilicate Glass
0.0081 in.
0.206 mm
0.0308 in.
7.82 mm
2.75 in.
239°F
50°F
10°C
2 μL
Sharp, Bevel, Curved, Non-coring
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers