missing translation for 'onlineSavingsMsg'
Learn More
2-Hydroxybenzonitrile, 98%, Thermo Scientific Chemicals Product Code.: 11420164

2-Hydroxybenzonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11420164
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11420164

Brand: Thermo Scientific Alfa Aesar A13606.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Hydroxybenzonitrile is used as starting reagent during the synthesis of mono-alkoxyphenyloxazoline, furanoside.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxybenzonitrile is used as starting reagent during the synthesis of mono-alkoxyphenyloxazoline, furanoside.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-20-1
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.123
MDL Number MFCD00002145
InChI Key CHZCERSEMVWNHL-UHFFFAOYSA-N
Synonym 2-cyanophenol, o-cyanophenol, salicylonitrile, o-hydroxybenzonitrile, benzonitrile, 2-hydroxy, salicylnitrile, benzonitrile, hydroxy, benzonitrile, o-hydroxy, o-hydoxybenzonitrile, 2-hydroxy-benzonitrileShow MoreShow Less
PubChem CID 11907
IUPAC Name 2-hydroxybenzonitrile
SMILES C1=CC=C(C(=C1)C#N)O

Specifications

Melting Point 92°C to 95°C
Boiling Point 149°C (14 mmHg)
Quantity 25 g
Beilstein 1210029
Solubility Information Soluble in water.
Formula Weight 119.12
Percent Purity 98%
Chemical Name or Material 2-Hydroxybenzonitrile

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10220248 Certificate TypeCertificate of Analysis Date21/03/2024 Catalog NumberA13606.06,A13606.14,A13606.22
Lot Number10175780 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13606.06,A13606.14,A13606.22
Lot Number10184785 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13606.06,A13606.14,A13606.22
Lot Number10203264 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13606.06,A13606.14,A13606.22
Lot Number10129762 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13606.06,A13606.14,A13606.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Hydroxybenzonitrile, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.