missing translation for 'onlineSavingsMsg'
Learn More
2-Fluoro-5-methoxybenzaldehyde, 97%, Thermo Scientific Chemicals Product Code.: 11483148

2-Fluoro-5-methoxybenzaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 11483148
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11483148

Brand: Thermo Scientific Alfa Aesar A18904.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105728-90-3
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00070795
InChI Key DKIQXHIAEMGZGO-UHFFFAOYSA-N
Synonym 6-fluoro-m-anisaldehyde, 2-fluoro-5-methoxy-benzaldehyde, 2-fluoro-5-methoxy benzaldehyde, benzaldehyde, 2-fluoro-5-methoxy, pubchem2634, 4-fluoro-3-formylanisole, acmc-2098hi, ksc490m2b, 2-fluoro-5-methyloxy benzaldehyde, 2-fluoro-5-methoxybenzaldehyde
PubChem CID 2734872
IUPAC Name 2-fluoro-5-methoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)F)C=O

Specifications

Density 1.237
Boiling Point 80°C (0.9 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.533
Quantity 5 g
Beilstein 4379088
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 154.14
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-methoxybenzaldehyde
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247574 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberA18904.06,A18904.03,A18904.14
Lot Number10109037 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18904.06,A18904.14
Lot Number10113677 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18904.06,A18904.14
Lot Number10135996 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18904.06,A18904.14
Lot Number10161897 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18904.06,A18904.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluoro-5-methoxybenzaldehyde, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.