missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoromandelic acid, 96%, Thermo Scientific™

Product Code. 11338305
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207853-61-0
Molecular Formula C8H6F2O3
Molecular Weight (g/mol) 188.13
MDL Number MFCD00061297
InChI Key PATKDMCCMCSATF-UHFFFAOYNA-N
Synonym 2,5-difluoromandelic acid, 2-2,5-difluorophenyl-2-hydroxyacetic acid, 2,5-difluorophenyl hydroxy acetic acid, 2,5-difluoro mandelic acid, 2,5-difluorophenyl hydroxy acetic acid #, 2-2,5-difluorophenyl-2-hydroxy-acetic acid, 2-2,5-bis fluoranyl phenyl-2-oxidanyl-ethanoic acid
PubChem CID 588649
IUPAC Name 2-(2,5-difluorophenyl)-2-hydroxyacetic acid
SMILES OC(C(O)=O)C1=C(F)C=CC(F)=C1

Specifications

Melting Point 110°C to 113°C
Quantity 1 g
Formula Weight 188.13
Percent Purity 96%
Chemical Name or Material 2,5-Difluoromandelic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'