missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoroaniline, 99%, Thermo Scientific Chemicals

Product Code. 11410284
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,4-Difluoroaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoroaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 367-25-9
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007648
InChI Key CEPCPXLLFXPZGW-UHFFFAOYSA-N
Synonym 2,4-difluorobenzenamine, benzenamine, 2,4-difluoro, 2,4-difluroaniline, 2,4-difluorophenylamine, aniline, 2,4-difluoro, unii-40p93l7kwd, 1-amino-2,4-difluorobenzene, ccris 4621, 2,4-difluoro aniline, 2,4-difluoro-phenylamine
PubChem CID 9709
IUPAC Name 2,4-difluoroaniline
SMILES NC1=CC=C(F)C=C1F

Spécification

Melting Point -8°C to -7°C
Density 1.281
Boiling Point 168°C to 170°C
Flash Point 62°C (143°F)
Odor Characteristic
Refractive Index 1.507
Quantity 500 g
UN Number UN2941
Beilstein 2802556
Merck Index 14,3147
Solubility Information Not miscible or difficult to mix.
Formula Weight 129.11
Percent Purity 99%
Chemical Name or Material 2,4-Difluoroaniline
Afficher plus Afficher moins

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'