Learn More
2,4,6-Trimethylstyrene, 95%, stab. with 500ppm 4-tert-butylcatechol, Thermo Scientific Chemicals
€ 27.03 - € 301.50
Chemical Identifiers
CAS 769-25-5
Molecular Formula C11H14
Molecular Weight (g/mol) 146.233
MDL Number MFCD00008613
InChI Key PDELBHCVXBSVPJ-UHFFFAOYSA-N
Synonym 2,4,6-trimethylstyrene, 1,3,5-trimethyl-2-vinylbenzene, 2-vinylmesitylene, mesitylethylene, vinylmesitylene, styrene, 2,4,6-trimethyl, unii-89fv7yq871, benzene, 2-ethenyl-1,3,5-trimethyl, 2,6-trimethylstyrene, styrene,4,6-trimethyl
PubChem CID 13036
IUPAC Name 2-ethenyl-1,3,5-trimethylbenzene
SMILES CC1=CC(=C(C(=C1)C)C=C)C
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11484090
SDS View Documents
Thermo Scientific Alfa Aesar
A15974.03
1 g
€ 27.03
1g
11448546
SDS View Documents
Thermo Scientific Alfa Aesar
A15974.06
5 g
€ 85.32
5g
11458546
SDS View Documents
Thermo Scientific Alfa Aesar
A15974.14
25 g
€ 301.50
25g
Description
Description
2,4,6-Trimethylstyrene is used as an intermediate to produce other chemicals like 2-Methoxy-1-(2, 4, 6-trimethyl-phenyl)-ethanone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4,6-Trimethylstyrene is used as an intermediate to produce other chemicals like 2-Methoxy-1-(2, 4, 6-trimethyl-phenyl)-ethanone.
Solubility
Soluble in methanol.
Notes
It is sensitive to light. Incompatible with oxidizing agents.
Specifications
Show More
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 212-205-4
missing translation for 'tsca' : No
Recommended Storage : Keep cold
SDS
Documents
Documents
Product Certifications
RUO – Research Use Only
Product Content Correction
Product Title
2,4,6-Trimethylstyrene, 95%, stab. with 500ppm 4-tert-butylcatechol, Thermo Scientific Chemicals