missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-2-(trifluoromethylsulfonyl)benzene, 95%, Thermo Scientific™

Product Code. 15475578
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2358-41-0
Molecular Formula C7H4F4O2S
Molecular Weight (g/mol) 228.161
MDL Number MFCD09971596
InChI Key NXZFXDLCZKKNCX-UHFFFAOYSA-N
Synonym 1-fluoro-2-trifluoromethyl sulfonyl benzene, 1-fluoro-2-trifluoromethylsulfonyl benzene, 1-fluoro-2-trifluoromethanesulfonylbenzene, o-fluorophenyl trifluoromethyl sulfone, 2-fluorophenyl trifluoromethyl sulfone, 1-fluoro-2-trifluoromethylsulfonyl-benzene, 1-fluoro-2-trifluoromethane sulfonylbenzene, 1-fluoro-2-trifluoromethanesulfonyl benzene, 1-fluoro-2-trifluoromethyl ;sulfonyl ;benzene
PubChem CID 15731661
IUPAC Name 1-fluoro-2-(trifluoromethylsulfonyl)benzene
SMILES C1=CC=C(C(=C1)F)S(=O)(=O)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 228.16
Percent Purity 95%
Chemical Name or Material 1-Fluoro-2-(trifluoromethylsulfonyl)benzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Fluoro-2-(trifluoromethylsulfonyl)benzene, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.