missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2,4-dinitrobenzene, 98%, Thermo Scientific Chemicals

€ 28.04 - € 319.59

Chemical Identifiers

CAS 97-00-7
Molecular Formula C6H3ClN2O4
Molecular Weight (g/mol) 202.55
MDL Number MFCD00007075
InChI Key VYZAHLCBVHPDDF-UHFFFAOYSA-N
Synonym 2,4-dinitrochlorobenzene, dinitrochlorobenzene, dncb, cdnb, chlorodinitrobenzene, 4-chloro-1,3-dinitrobenzene, 2,4-dinitrophenyl chloride, benzene, 1-chloro-2,4-dinitro, dinitrochlorobenzol, 1,3-dinitro-4-chlorobenzeneShow MoreShow Less
PubChem CID 6
ChEBI CHEBI:34718
IUPAC Name 1-chloro-2,4-dinitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11434834
SDS View Documents
Thermo Scientific Alfa Aesar
A13774.22
100 g
€ 28.04
100g
Please to purchase this item. Need a web account? Register with us today!
11444834
SDS View Documents
Thermo Scientific Alfa Aesar
A13774.36
500 g
€ 92.66
500g
Please to purchase this item. Need a web account? Register with us today!
11454834
SDS View Documents
Thermo Scientific Alfa Aesar
A13774.0E
2500 g
€ 319.59
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Chloro-2,4-dinitrobenzene is used as a reagent for the detection and determination of pyridine compounds. It has been used as alkylating agent to evaluate the depletion of intracellular erythrocyte glutathione (GSH). It is an irreversible inhibitor of human thioredoxin reductase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2,4-dinitrobenzene is used as a reagent for the detection and determination of pyridine compounds. It has been used as alkylating agent to evaluate the depletion of intracellular erythrocyte glutathione (GSH). It is an irreversible inhibitor of human thioredoxin reductase.

Solubility
Soluble in chloroform, ether, hot alcohol, and benzene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, ammonia. Reacts with hydrazine hydrate.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Chloro-2,4-dinitrobenzene, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.