Learn More
1-Chloro-2,4-dinitrobenzene, 98%, Thermo Scientific Chemicals
Description
1-Chloro-2,4-dinitrobenzene is used as a reagent for the detection and determination of pyridine compounds. It has been used as alkylating agent to evaluate the depletion of intracellular erythrocyte glutathione (GSH). It is an irreversible inhibitor of human thioredoxin reductase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Chloro-2,4-dinitrobenzene is used as a reagent for the detection and determination of pyridine compounds. It has been used as alkylating agent to evaluate the depletion of intracellular erythrocyte glutathione (GSH). It is an irreversible inhibitor of human thioredoxin reductase.
Solubility
Soluble in chloroform, ether, hot alcohol, and benzene. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, ammonia. Reacts with hydrazine hydrate.

Specifications
Specifications
Melting Point | 48°C to 52°C |
Density | 1.7 |
Boiling Point | 314°C to 316°C |
Flash Point | 186°C (366°F) |
Quantity | 500 g |
UN Number | UN3441 |
Beilstein | 613161 |
Merck Index | 14,2136 |
Solubility Information | Soluble in chloroform,ether,hot alcohol,and benzene. Insoluble in water. |
Formula Weight | 202.55 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10252959 | Certificate of Analysis | 05/11/2024 | A13774.36,A13774.0E,A13774.22 |
10169855 | Certificate of Analysis | 22/11/2021 | A13774.36,A13774.0E,A13774.22 |
10188717 | Certificate of Analysis | 22/11/2021 | A13774.36,A13774.0E,A13774.22 |
10213299 | Certificate of Analysis | 22/11/2021 | A13774.36,A13774.0E,A13774.22 |
10163974 | Certificate of Analysis | 22/11/2021 | A13774.36,A13774.0E,A13774.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.