missing translation for 'onlineSavingsMsg'
Learn More
1-Bromo-4-fluorobenzene, 99%, Thermo Scientific Chemicals Product Code.: 11404733

1-Bromo-4-fluorobenzene, 99%, Thermo Scientific Chemicals

Product Code. 11404733
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11404733

Brand: Thermo Scientific Alfa Aesar A11630.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Bromo-4-fluorobenzene is used as an intermediate in the synthesis of atypical antipsychotic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-fluorobenzene is used as an intermediate in the synthesis of atypical antipsychotic agents.

Solubility
Soluble in water(0.14g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 460-00-4
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175.00
MDL Number MFCD00000342
InChI Key AITNMTXHTIIIBB-UHFFFAOYSA-N
Synonym 4-bromofluorobenzene, p-bromofluorobenzene, p-fluorobromobenzene, 4-fluorobromobenzene, benzene, 1-bromo-4-fluoro, 4-fluorophenyl bromide, 1-fluoro-4-bromobenzene, 4-fluoro-1-bromobenzene, p-fluorophenyl bromide, bromo fluorobenzeneShow MoreShow Less
PubChem CID 9993
IUPAC Name 1-bromo-4-fluorobenzene
SMILES FC1=CC=C(Br)C=C1

Specifications

Melting Point -16°C
Density 1.604
Boiling Point 151°C to 153°C
Flash Point 60°C (140°F)
Odor Aromatic
Refractive Index 1.527
Quantity 500 g
UN Number UN1993
Beilstein 774102
Solubility Information Soluble in water(0.14g/L).
Formula Weight 175.01
Percent Purity 99%
Chemical Name or Material 1-Bromo-4-fluorobenzene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242211 Certificate TypeCertificate of Analysis Date20/07/2022 Catalog NumberA11630.0E,A11630.36,A11630.22
Lot Number10238958 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA11630.0E,A11630.36,A11630.22
Lot Number10200744 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11630.0E,A11630.36,A11630.22
Lot Number10139671 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11630.0E,A11630.36,A11630.22
Lot Number10155129 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11630.0E,A11630.36,A11630.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromo-4-fluorobenzene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.