missing translation for 'onlineSavingsMsg'
Learn More
1-Bromo-3-ethylbenzene, 98%, Thermo Scientific Chemicals Product Code.: 11404963

1-Bromo-3-ethylbenzene, 98%, Thermo Scientific Chemicals

Product Code. 11404963
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11404963

Brand: Thermo Scientific Alfa Aesar A12258.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2725-82-8
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00156128
InChI Key ZRFJYAZQMFCUIX-UHFFFAOYSA-N
Synonym benzene, 1-bromo-3-ethyl, 3-ethylbromobenzene, 1-bromo-3-ethyl-benzene, 3-bromoethyl benzene, 3ethylbromobenzene, m-bromoethylbenzene, 3-bromo-ethylbenzene, pubchem3771, 1bromo-3-ethylbenzene, 1-bromo-3-ethylbenzeShow MoreShow Less
PubChem CID 123170
IUPAC Name 1-bromo-3-ethylbenzene
SMILES CCC1=CC(=CC=C1)Br

Specifications

Density 1.3493
Boiling Point 76°C to 78°C (10 mmHg)
Quantity 5 g
Formula Weight 185.06
Percent Purity 98%
Chemical Name or Material 1-Bromo-3-ethylbenzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10193290 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA12258.03,A12258.14,A12258.06
Lot NumberI3644A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12258.03,A12258.14,A12258.06
Lot Number10193290 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12258.03,A12258.14,A12258.06
Lot NumberI3396A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12258.03,A12258.14,A12258.06
Lot Number10193290 Certificate TypeCertificate of Analysis Date06/08/2020 Catalog NumberA12258.03,A12258.14,A12258.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromo-3-ethylbenzene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.