missing translation for 'onlineSavingsMsg'
Learn More
1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene, 98%, Thermo Scientific™ Product Code.: 15498338

1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene, 98%, Thermo Scientific™

Product Code. 15498338
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15498338

Brand: Thermo Scientific Alfa Aesar H61771.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168971-68-4
Molecular Formula C7H3BrF4O
Molecular Weight (g/mol) 258.998
MDL Number MFCD06657548
InChI Key KZMHSGBETSENAT-UHFFFAOYSA-N
Synonym 1-bromo-2-fluoro-4-trifluoromethoxy benzene, 4-bromo-3-fluoro trifluoromethoxy benzene, 3-fluoro-4-bromobenzenetrifluoromethoxy, 4-bromo-3-fluoro-trifluoromethoxy benzene, benzene, 1-bromo-2-fluoro-4-trifluoromethoxy, pubchem1075, acmc-1c50f, ksc498c3b, 4-bromo-3-fluorotrifluoromethoxy-benzene, 2-fluoro-4-trifluoromethoxy bromobenzeneShow MoreShow Less
PubChem CID 22065211
IUPAC Name 1-bromo-2-fluoro-4-(trifluoromethoxy)benzene
SMILES C1=CC(=C(C=C1OC(F)(F)F)F)Br

Specifications

Color Yellow
Quantity 1 g
Formula Weight 259
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81400815 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number81301395 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.