missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenylsulfonyl)-3,3-dimethyl-2-butanone, 98+%, Thermo Scientific™

Product Code. 11317766
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207974-06-9
Molecular Formula C12H15ClO3S
Molecular Weight (g/mol) 274.759
MDL Number MFCD00082685
InChI Key XTIUXIMZIKBBSU-UHFFFAOYSA-N
Synonym 1-4-chlorobenzenesulfonyl-3,3-dimethylbutan-2-one, 1-4-chlorophenyl sulfonyl-3,3-dimethylbutan-2-one, 1-4'-chlorobenzenesulfonyl-3,3-dimethylbutane-2-one, 1-4-chlorophenylsulfonyl-3,3-dimethyl-2-butanone, 1-4-chlorophenyl sulfonyl-3,3-dimethyl-2-butanone, 1-4'-chlorophenylsulfonyl-3,3-dimethylbutane-2-one, 1-4-chlorobenzenesulfonyl-3,3-dimethyl-2-butanone, 1-4-chlorophenyl sulfonyl-3,3-dimethyl-butan-2-one
PubChem CID 545199
IUPAC Name 1-(4-chlorophenyl)sulfonyl-3,3-dimethylbutan-2-one
SMILES CC(C)(C)C(=O)CS(=O)(=O)C1=CC=C(C=C1)Cl

Specifications

Melting Point 95°C to 99°C
Quantity 5 g
Formula Weight 274.77
Percent Purity ≥98%
Chemical Name or Material 1-(4-Chlorophenylsulfonyl)-3,3-dimethyl-2-butanone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.