missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-methyl-5-nitrobenzene, 95%, Thermo Scientific™

€ 69.20 - € 496.80

Chemical Identifiers

CAS 7149-69-1
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.022
MDL Number MFCD00174267
InChI Key RUVCGWLHTVGNGI-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-nitrotoluene, 1,3-dichloro-2-methyl-5-nitro-benzene, benzene, 1,3-dichloro-2-methyl-5-nitro, pubchem9506, maybridge1_005049, ksc496m1j
PubChem CID 251670
IUPAC Name 1,3-dichloro-2-methyl-5-nitrobenzene
SMILES CC1=C(C=C(C=C1Cl)[N+](=O)[O-])Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10496662
View Documents
Thermo Scientific Maybridge
KM08058DA
1 g Amber Glass Bottle
€ 69.20
1g
Please to purchase this item. Need a web account? Register with us today!
10692063
View Documents
Thermo Scientific Maybridge
KM08058EA
10 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

7149-69-1
206.022
RUVCGWLHTVGNGI-UHFFFAOYSA-N
251670
CC1=C(C=C(C=C1Cl)[N+](=O)[O-])Cl
C7H5Cl2NO2
MFCD00174267
2,6-dichloro-4-nitrotoluene, 1,3-dichloro-2-methyl-5-nitro-benzene, benzene, 1,3-dichloro-2-methyl-5-nitro, pubchem9506, maybridge1_005049, ksc496m1j
1,3-dichloro-2-methyl-5-nitrobenzene

Specifications

7149-69-1
95%
MFCD00174267
RUVCGWLHTVGNGI-UHFFFAOYSA-N
1,3-dichloro-2-methyl-5-nitrobenzene
251670
95%
95.0
C7H5Cl2NO2
2,6-dichloro-4-nitrotoluene, 1,3-dichloro-2-methyl-5-nitro-benzene, benzene, 1,3-dichloro-2-methyl-5-nitro, pubchem9506, maybridge1_005049, ksc496m1j
CC1=C(C=C(C=C1Cl)[N+](=O)[O-])Cl
206.022
206.03
1, 3-Dichloro-2-methyl-5-nitrobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.