missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
(1,1,2,2-Tetrafluoroethoxy)benzene, 98%, Thermo Scientific™
Klik for at se tilgængelige muligheder
Quantity:
1 g
5 g
25 g
Pakningsstørrelse:
1g
25g
5g
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Tekniske data
Tekniske data
| Density | 1.28 |
| Boiling Point | 152°C to 153°C |
| Flash Point | 46°C (115°F) |
| Refractive Index | 1.421 |
| Quantity | 25 g |
| UN Number | UN1993 |
| Formula Weight | 194.13 |
| Percent Purity | 98% |
| Chemical Name or Material | (1,1,2,2-Tetrafluoroethoxy)benzene |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion