accessibility menu, dialog, popup

UserName

CAS RN 870837-66-4

CH3OFOF

CAS RN 870837-66-4

IUPAC Name: 2,4-difluoro-3-methoxybenzaldehyde
Synonyms: 2,4-difluoro-3-methoxybenzaldehyde
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: CEEXKEWRNDNTKK-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(C=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results