CAS RN 85-36-9
CAS RN 85-36-9
IUPAC Name:
3-acetamido-2,4,6-triiodobenzoic acid
Synonyms:
acetrizoic acid
Molecular Weight (g/mol):
556.86
Molecular Formula:
C9H6I3NO3
InChi Key:
GNOGSFBXBWBTIG-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=C(I)C=C(I)C(C(O)=O)=C1I
1
–
1
of
1
results
1
Acetrizoic acid, MedChemExpress
MedChemExpress Acetrizoic acid is a molecule used as a contrast medium.