CAS RN 80573-04-2
CAS RN 80573-04-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Balsalazide, MedChemExpress
MedChemExpress Balsalazide could suppress colitis-associated carcinogenesis through modulation of IL-6/STAT3 pathway.