CAS RN 31637-97-5
CAS RN 31637-97-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Etofibrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Etofibrate, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
3
Etofibrate, MedChemExpress
MedChemExpress Etofibrate is the ethandiol-1,2 diester of the nicotinic and clofibric acids. Etofibrate has been shown to be a potent hypolipidemic agent in animal and human.