man

Test

CAS RN 102-92-1

ClO

CAS RN 102-92-1

IUPAC Name: (2E)-3-phenylprop-2-enoyl chloride
Synonyms: (2E)-3-phenylprop-2-enoyl chloride
Molecular Weight (g/mol): 166.6
Molecular Formula: C9H7ClO
InChi Key: WOGITNXCNOTRLK-VOTSOKGWSA-N
SMILES: ClC(=O)\C=C\C1=CC=CC=C1
Boiling Point 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)