man

Test

CAS RN 102-56-7

H3COOCH3H2N

CAS RN 102-56-7

IUPAC Name: 2,5-dimethoxyaniline
Synonyms: 2,5-dimethoxyaniline
Molecular Weight (g/mol): 153.18
Molecular Formula: C8H11NO2
InChi Key: NAZDVUBIEPVUKE-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(N)=C1
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)