man

Test

CAS RN 102-47-6

ClClCl

CAS RN 102-47-6

IUPAC Name: 1,2-dichloro-4-(chloromethyl)benzene
Synonyms: 1,2-dichloro-4-(chloromethyl)benzene
Molecular Weight (g/mol): 195.47
Molecular Formula: C7H5Cl3
InChi Key: YZIFVWOCPGPNHB-UHFFFAOYSA-N
SMILES: ClCC1=CC=C(Cl)C(Cl)=C1
Boiling Point 
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (4)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (4)
  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)