CAS RN 10075-85-1
CAS RN 10075-85-1
IUPAC Name:
9,10-bis(2-phenylethynyl)anthracene
Synonyms:
Bis(PhCC)-Anthracene
Molecular Weight (g/mol):
378.47
Molecular Formula:
C30H18
InChi Key:
ZHBOFZNNPZNWGB-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)C#CC1=C2C=CC=CC2=C(C#CC2=CC=CC=C2)C2=CC=CC=C12
1
–
1
of
1
results
1
9,10-Bis(phenylethynyl)anthracene, Thermo Scientific Chemicals
CAS: 10075-85-1 Molecular Formula: C30H18 Molecular Weight (g/mol): 378.474 MDL Number: MFCD00012050 InChI Key: ZHBOFZNNPZNWGB-UHFFFAOYSA-N Synonym: 9,10-bis phenylethynyl anthracene,9,10-bis 2-phenylethynyl anthracene,anthracene, 9,10-bis phenylethynyl,bpea,anthracene, 9,10-bis 2-phenylethynyl,bis phcc-anthracene,unii-fc8jdb70dq,9,10-bis phenylvinyl anthracene,fc8jdb70dq,9,10-bis-phenylethynyl anthracene PubChem CID: 82338 ChEBI: CHEBI:51675 IUPAC Name: 9,10-bis(2-phenylethynyl)anthracene SMILES: C1=CC=C(C=C1)C#CC2=C3C=CC=CC3=C(C4=CC=CC=C42)C#CC5=CC=CC=C5