man

Test

CAS RN 1003-99-2

NH2FBr

CAS RN 1003-99-2

IUPAC Name: 2-bromo-5-fluoroaniline
Synonyms: 2-bromo-5-fluoroaniline
Molecular Weight (g/mol): 190.02
Molecular Formula: C6H5BrFN
InChi Key: FWTXFEKVHSFTDQ-UHFFFAOYSA-N
SMILES: NC1=CC(F)=CC=C1Br
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)